siRNA Details
| Gene Symbol: Otor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020595.2 | NP_065620.1 | 1, 2 | 121 | ||
| GenBank mRNA | AF243504.1 | - | 1, 2 | 121 | ||
| AJ243939.1 | - | 1, 2 | 136 | |||
| BC139152.1 | - | 1, 2 | 220 | 8862174 | ||
| BC139153.1 | - | 1, 2 | 220 | 8862175 | ||

Otor
Genomic Map