siRNA Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020606.5 | NP_065631.3 | 13 | 2956 | ||
| GenBank mRNA | AB045321.1 | - | 13 | 2829 | ||
| AF237774.1 | - | 13 | 2829 | |||
| AK008197.1 | - | 1 | 347 | |||
| BC059236.1 | - | 13 | 2956 | 6400393 | ||

Parva
Genomic Map