siRNA Details
| Gene Symbol: BC051019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040700.2 | NP_001035790.1 | 4 | 793 | ||
| GenBank mRNA | AK143006.1 | - | 4 | 481 | ||
| BC051019.1 | - | 4 | 793 | 6390147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC051019| Gene Symbol: BC051019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040700.2 | NP_001035790.1 | 4 | 793 | ||
| GenBank mRNA | AK143006.1 | - | 4 | 481 | ||
| BC051019.1 | - | 4 | 793 | 6390147 | ||