siRNA Details
| Gene Symbol: Akip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020616.1 | NP_065641.1 | 4 | 388 | ||
| GenBank mRNA | AK086219.1 | - | 4 | 421 | ||
| AK155010.1 | - | 4 | 437 | |||
| AK159903.1 | - | 4 | 389 | |||
| AK163012.1 | - | 4 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akip1| Gene Symbol: Akip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020616.1 | NP_065641.1 | 4 | 388 | ||
| GenBank mRNA | AK086219.1 | - | 4 | 421 | ||
| AK155010.1 | - | 4 | 437 | |||
| AK159903.1 | - | 4 | 389 | |||
| AK163012.1 | - | 4 | 435 | |||