siRNA Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021303.2 | NP_067278.2 | 17 | 2053 | ||
| GenBank mRNA | AF155546.1 | - | 5 | 563 | ||
| AK076287.1 | - | 16 | 2095 | |||
| AK133834.1 | - | 17 | 2053 | |||
| AK157765.1 | - | 17 | 2046 | |||
| AK199140.1 | - | 1 | 56 | |||
| BC020013.1 | - | 17 | 2024 | 3663482 | ||

Noc2l
Genomic Map