siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021305.3 | NP_067280.1 | 12 | 1902 | ||
| GenBank mRNA | AK017615.1 | - | 11 | 1686 | ||
| AK077701.1 | - | 12 | 1902 | |||
| AK160602.1 | - | 12 | 1886 | |||
| BC005458.1 | - | 12 | 1845 | 3494001 | ||

Sec61a2
Genomic Map