siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021305.3 | NP_067280.1 | 12 | 2252 | ||
| GenBank mRNA | AK017615.1 | - | 11 | 2036 | ||
| AK077701.1 | - | 12 | 2252 | |||
| AK160602.1 | - | 12 | 2236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec61a2| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021305.3 | NP_067280.1 | 12 | 2252 | ||
| GenBank mRNA | AK017615.1 | - | 11 | 2036 | ||
| AK077701.1 | - | 12 | 2252 | |||
| AK160602.1 | - | 12 | 2236 | |||