siRNA Details
| Gene Symbol: Cend1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021316.4 | NP_067291.1 | 2 | 705 | ||
| GenBank mRNA | AF243130.1 | - | 2 | 643 | ||
| AK005092.1 | - | 2 | 680 | |||
| AK079045.1 | - | 2 | 706 | |||
| BC023032.1 | - | 2 | 643 | 5366891 | ||

Cend1
Genomic Map