siRNA Details
| Gene Symbol: Fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021318.3 | NP_067293.1 | 3 | 477 | ||
| GenBank mRNA | AF083394.1 | - | 3 | 425 | ||
| AK006414.1 | - | 3 | 466 | |||
| BC048473.1 | - | 3 | 477 | 6770515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhl5| Gene Symbol: Fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021318.3 | NP_067293.1 | 3 | 477 | ||
| GenBank mRNA | AF083394.1 | - | 3 | 425 | ||
| AK006414.1 | - | 3 | 466 | |||
| BC048473.1 | - | 3 | 477 | 6770515 | ||