siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301692.1 | NP_001288621.1 | 6 | 1023 | ||
| NM_001301694.1 | NP_001288623.1 | 6 | 973 | |||
| NM_021340.4 | NP_067315.1 | 7 | 1145 | |||
| GenBank mRNA | AF076930.1 | - | 7 | 1065 | ||
| AK044450.1 | - | 7 | 1097 | |||
| AK044649.1 | - | 7 | 1096 | |||
| AK149272.1 | - | 7 | 1096 | |||
| BC082611.1 | - | 7 | 1082 | |||
| BC086689.1 | - | 6 | 1559 | |||
More Information
Set Membership: |
|

Rgr
Genomic Map