siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301692.1 | NP_001288621.1 | 6 | 1494 | ||
| NM_001301694.1 | NP_001288623.1 | 6 | 1444 | |||
| NM_021340.4 | NP_067315.1 | 7 | 1616 | |||
| GenBank mRNA | AK044450.1 | - | 7 | 1568 | ||
| AK044649.1 | - | 7 | 1567 | |||
| BC082611.1 | - | 7 | 1553 | |||
| BC086689.1 | - | 6 | 2030 | |||
More Information
Set Membership: |
|

Rgr
Genomic Map