siRNA Details
| Gene Symbol: Extl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163514.1 | NP_001156986.1 | 3 | 550 | ||
| NM_001163515.1 | NP_001156987.1 | 2 | 668 | |||
| NM_021388.4 | NP_067363.3 | 3 | 743 | |||
| GenBank mRNA | AB032170.1 | - | 3 | 553 | ||
| AB032171.1 | - | 3 | 538 | |||
| AF200973.1 | - | 3 | 515 | |||
| AK019370.1 | - | 3 | 481 | |||
| AK028677.1 | - | 3 | 716 | |||
| AK032016.1 | - | 1 | 76 | |||
| AK159281.1 | - | 3 | 730 | |||
| AK161866.1 | - | 2 | 634 | |||
| BC031438.1 | - | 3 | 707 | 4024555 | ||
| BC094444.1 | - | 3 | 456 | 4507232 | ||

Extl2
Genomic Map