siRNA Details
| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021391.3 | NP_067366.1 | 7 | 822 | ||
| GenBank mRNA | AK002813.1 | - | 3 | 456 | ||
| BC025123.1 | - | 7 | 791 | 4019442 | ||
| W75893.1 | - | 2 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r1a| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021391.3 | NP_067366.1 | 7 | 822 | ||
| GenBank mRNA | AK002813.1 | - | 3 | 456 | ||
| BC025123.1 | - | 7 | 791 | 4019442 | ||
| W75893.1 | - | 2 | 232 | |||