siRNA Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021422.4 | NP_067397.1 | 2 | 467 | ||
| GenBank mRNA | AB032401.1 | - | 3 | 334 | ||
| AK016666.1 | - | 1 | 64 | |||
| AK046476.1 | - | 2 | 468 | |||
| AK076175.1 | - | 2 | 468 | |||
| AK141909.1 | - | 2 | 469 | |||
| AK197546.1 | - | 2 | 199 | |||
| BC147484.1 | - | 2 | 315 | 9056231 | ||
| BC147486.1 | - | 2 | 315 | 9056239 | ||

Dnaja4
Genomic Map