siRNA Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021422.4 | NP_067397.1 | 5 | 832 | ||
| GenBank mRNA | AB032401.1 | - | 6 | 699 | ||
| AK016666.1 | - | 4 | 429 | |||
| AK046476.1 | - | 5 | 833 | |||
| AK076175.1 | - | 5 | 833 | |||
| AK141909.1 | - | 5 | 834 | |||
| BC022948.1 | - | 3 | 330 | |||
| BC147484.1 | - | 5 | 680 | 9056231 | ||
| BC147486.1 | - | 5 | 680 | 9056239 | ||

Dnaja4
Genomic Map