siRNA Details
| Gene Symbol: Pvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021424.2 | NP_067399.2 | 6 | 2159 | ||
| GenBank mRNA | AK147479.1 | - | 2752 | |||
| BC060694.1 | - | 6 | 2159 | 6405088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl1| Gene Symbol: Pvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021424.2 | NP_067399.2 | 6 | 2159 | ||
| GenBank mRNA | AK147479.1 | - | 2752 | |||
| BC060694.1 | - | 6 | 2159 | 6405088 | ||