siRNA Details
| Gene Symbol: Chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021439.2 | NP_067414.2 | 3 | 1580 | ||
| GenBank mRNA | AJ289133.1 | - | 3 | 1588 | ||
| BC137629.1 | - | 3 | 1332 | 8860648 | ||
| BC144793.1 | - | 3 | 1313 | 9053341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst11| Gene Symbol: Chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021439.2 | NP_067414.2 | 3 | 1580 | ||
| GenBank mRNA | AJ289133.1 | - | 3 | 1588 | ||
| BC137629.1 | - | 3 | 1332 | 8860648 | ||
| BC144793.1 | - | 3 | 1313 | 9053341 | ||