siRNA Details
| Gene Symbol: Chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021439.2 | NP_067414.2 | 3 | 724 | ||
| GenBank mRNA | AB030378.1 | - | 3 | 876 | ||
| AJ289133.1 | - | 3 | 732 | |||
| BC137629.1 | - | 3 | 472 | 8860648 | ||
| BC144793.1 | - | 3 | 457 | 9053341 | ||

Chst11
Genomic Map