siRNA Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021504.3 | NP_067479.2 | 3 | 315 | ||
| GenBank mRNA | AF250927.1 | - | 3 | 302 | ||
| AK028248.1 | - | 3 | 316 | |||
| AK148931.1 | - | 3 | 318 | |||
| AK169205.1 | - | 3 | 303 | |||
| BC028961.1 | - | 3 | 269 | 5366023 | ||

Ngly1
Genomic Map