siRNA Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021504.3 | NP_067479.2 | 5, 6 | 922 | ||
| GenBank mRNA | AF250927.1 | - | 5, 6 | 909 | ||
| AK028248.1 | - | 5, 6 | 923 | |||
| AK148931.1 | - | 5, 6 | 925 | |||
| AK169205.1 | - | 5, 6 | 910 | |||
| BC028961.1 | - | 5, 6 | 876 | 5366023 | ||

Ngly1
Genomic Map