siRNA Details
| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021512.2 | NP_067487.1 | 24, 25 | 3452 | ||
| GenBank mRNA | AF104415.2 | - | 24, 25 | 3452 | ||
| AK155492.1 | - | 23, 24 | 2826 | |||
| AK172911.1 | - | 10, 11 | 1178 | |||
| BC052450.1 | - | 23, 24 | 2810 | 6409240 | ||
| BC054523.1 | - | 23, 24 | 2810 | 6409240 | ||

Nup160
Genomic Map