siRNA Details
| Gene Symbol: Rab2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021518.3 | NP_067493.1 | 8 | 1574 | ||
| GenBank mRNA | AK042862.1 | - | 8 | 1544 | ||
| AK079071.1 | - | 8 | 1575 | |||
| AK135421.1 | - | 8 | 1558 | |||
| AK146179.1 | - | 8 | 1559 | |||
| AK150030.1 | - | 8 | 1606 | |||
| AK159607.1 | - | 8 | 1595 | |||
| AK160333.1 | - | 8 | 1575 | |||
| AK163320.1 | - | 8 | 1558 | |||

Rab2a
Genomic Map