siRNA Details
| Gene Symbol: Chst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021528.3 | NP_067503.3 | 2 | 406 | ||
| GenBank mRNA | AJ289132.1 | - | 1 | 319 | ||
| AK142155.1 | - | 2 | 404 | |||
| AK163588.1 | - | 2 | 376 | |||
| AK170209.1 | - | 2 | 369 | |||
| AK170484.1 | - | 2 | 373 | |||
| AK171208.1 | - | 2 | 373 | |||
| BC003201.1 | - | 2 | 387 | 3586766 | ||

Chst12
Genomic Map