siRNA Details
| Gene Symbol: Mettl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021554.2 | NP_067529.2 | 1, 2 | 536 | ||
| GenBank mRNA | AB041664.1 | - | 1, 2 | 314 | ||
| AJ278576.1 | - | 1, 2 | 536 | |||
| AK075586.1 | - | 1, 2 | 299 | |||
| AK201397.1 | - | 1, 2 | 24 | |||
| BC068124.1 | - | 1, 2 | 330 | 5716972 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001163164, XM_008759658, XM_006230154, HQ898855.

Mettl9
Genomic Map