siRNA Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB036882.1 | - | 9 | 2338 | ||
| AK154535.1 | - | 8 | 2279 | |||
| AK164324.1 | - | 8 | 2227 | |||
| AK164651.1 | - | 8 | 2226 | |||
| BC034719.1 | - | 9 | 2308 | 3710931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Midn| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB036882.1 | - | 9 | 2338 | ||
| AK154535.1 | - | 8 | 2279 | |||
| AK164324.1 | - | 8 | 2227 | |||
| AK164651.1 | - | 8 | 2226 | |||
| BC034719.1 | - | 9 | 2308 | 3710931 | ||