siRNA Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021605.3 | NP_067618.1 | 10 | 1768 | ||
| GenBank mRNA | AF217650.1 | - | 9 | 1622 | ||
| AK035502.1 | - | 10 | 1767 | |||
| AK088173.1 | - | 10 | 1817 | |||
| AK149308.1 | - | 10 | 1814 | |||
| AK165583.1 | - | 8 | 1537 | |||
| AK169649.1 | - | 1 | 604 | |||
| BC026541.1 | - | 10 | 1758 | |||
| BC037697.1 | - | 10 | 1746 | 3994871 | ||

Nek7
Genomic Map