siRNA Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159631.1 | NP_001153103.1 | 10 | 2049 | ||
| NM_021606.3 | NP_067619.1 | 10 | 2042 | |||
| GenBank mRNA | AF218847.1 | - | 10 | 2043 | ||
| AK004925.1 | - | 10 | 2019 | |||
| AK029266.1 | - | 10 | 2047 | |||
| AK075717.1 | - | 10 | 2015 | |||
| AK075836.1 | - | 10 | 2028 | |||
| AK086700.1 | - | 2047 | ||||
| AK089919.1 | - | 10 | 2002 | |||
| AK170757.1 | - | 10 | 2049 | |||
| AK208733.1 | - | 1 | 240 | |||

Nek6
Genomic Map