siRNA Details
| Gene Symbol: Cd274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021893.3 | NP_068693.1 | 7 | 2109 | ||
| GenBank mRNA | AF233517.1 | - | 6 | 2042 | ||
| AK154403.1 | - | 7 | 2088 | |||
| BC066841.1 | - | 7 | 2092 | 30101931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd274| Gene Symbol: Cd274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021893.3 | NP_068693.1 | 7 | 2109 | ||
| GenBank mRNA | AF233517.1 | - | 6 | 2042 | ||
| AK154403.1 | - | 7 | 2088 | |||
| BC066841.1 | - | 7 | 2092 | 30101931 | ||