siRNA Details
| Gene Symbol: Mapk8ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021921.3 | NP_068740.3 | 5 | 1987 | ||
| GenBank mRNA | AF220195.1 | - | 5 | 1706 | ||
| AF310135.1 | - | 5 | 1768 | |||
| AK014339.1 | - | 5 | 1691 | |||
| AK158968.1 | - | 5 | 1841 | |||
| BC029704.1 | - | 5 | 1766 | 5359447 | ||

Mapk8ip2
Genomic Map