siRNA Details
| Gene Symbol: Trpv4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022017.3 | NP_071300.2 | 8 | 1478 | ||
| GenBank mRNA | AB021875.2 | - | 7 | 1405 | ||
| AF208026.1 | - | 8 | 1446 | |||
| AF263522.1 | - | 7 | 1436 | |||
| AF279672.1 | - | 8 | 1462 | |||
| AJ296078.1 | - | 7 | 1397 | |||
| AK083223.1 | - | 8 | 1449 | |||
| AK158652.1 | - | 4 | 1331 | |||
| BC127052.1 | - | 8 | 1443 | 40109678 | ||

Trpv4
Genomic Map