siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022018.3 | NP_071301.2 | 8 | 1017 | ||
| GenBank mRNA | AB049355.1 | - | 8 | 857 | ||
| AK029847.1 | - | 4 | 329 | |||
| AK136597.1 | - | 8 | 927 | |||
| AK170193.1 | - | 8 | 940 | |||
| BC021332.1 | - | 8 | 905 | |||
| BC038006.1 | - | 8 | 873 | |||
| BC058234.1 | - | 6 | 617 | 6306849 | ||
| BC099690.1 | - | 8 | 924 | |||
| BC137843.1 | - | 4 | 394 | 8860862 | ||

Fam129a
Genomic Map