siRNA Details
| Gene Symbol: Ube4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022022.3 | NP_071305.2 | 2 | 852 | ||
| GenBank mRNA | AF260926.1 | - | 2 | 967 | ||
| AK016689.1 | - | 2 | 853 | |||
| AK029914.1 | - | 2 | 457 | |||
| AK083409.1 | - | 2 | 452 | |||
| AK122345.1 | - | 2 | 363 | |||
| BC067402.1 | - | 2 | 599 | 30099547 | ||
| BC075620.1 | - | 2 | 813 | 6405514 | ||

Ube4b
Genomic Map