siRNA Details
Gene Symbol: Smoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022315.2 | NP_071710.2 | 1, 2 | 301 | ||
GenBank mRNA | AJ249901.1 | - | 1, 2 | 272 | ||
AK030958.1 | - | 1, 2 | 270 | |||
AK040109.1 | - | 1, 2 | 249 | |||
AK159749.1 | - | 1, 2 | 268 | |||
AK160002.1 | - | 1, 2 | 268 | |||
AK172312.1 | - | 1, 2 | 266 | |||
BB840063.1 | - | 1, 2 | 302 | |||
BC019527.1 | - | 1, 2 | 248 | 4191849 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
