siRNA Details
| Gene Symbol: Gm20604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142939.1 | NP_001136411.1 | 2 | 1024 | ||
| GenBank mRNA | AK019599.1 | - | 1887 | |||
| BC014715.1 | - | 2 | 973 | 4236212 | ||
| BC055374.1 | - | 2 | 977 | 30071431 | ||
| Gene Symbol: Moap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142937.2 | NP_001136409.1 | 2 | 873 | ||
| NM_022323.7 | NP_071718.1 | 2 | 917 | |||
| GenBank mRNA | AF305551.1 | - | 1 | 648 | ||

Gm20604
Genomic Map