siRNA Details
| Gene Symbol: Sdf2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022324.3 | NP_071719.1 | 2 | 309 | ||
| GenBank mRNA | AB043006.1 | - | 2 | 249 | ||
| BC023290.1 | - | 2 | 263 | |||
| BC053425.1 | - | 2 | 291 | 6508268 | ||
| BC053438.1 | - | 2 | 291 | |||
| BY253823.1 | - | 2 | 310 | |||

Sdf2l1
Genomic Map