siRNA Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286133.1 | NP_001273062.1 | 2, 3 | 337 | ||
| NM_006638.3 | NP_006629.2 | 2, 3 | 337 | |||
| XM_011514253.2 | XP_011512555.1 | 2, 3 | 332 | |||
| GenBank mRNA | BG430780.1 | - | 2, 3 | 312 | ||
| BM015573.1 | - | 2, 3 | 369 | |||
| BM827175.1 | - | 2, 3 | 285 | |||
| BQ220155.1 | - | 2, 3 | 285 | |||

RPP40
Genomic Map