siRNA Details
Gene Symbol: Gpr88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022427.2 | NP_071872.1 | 2 | 1910 | ||
GenBank mRNA | AB042408.1 | - | 2 | 1880 | ||
BC051941.1 | - | 1 | 1526 | 5708059 | ||
BC157981.1 | - | 1 | 1519 | 9007300 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
