siRNA Details
| Gene Symbol: Ms4a8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022430.2 | NP_071875.2 | 7 | 911 | ||
| GenBank mRNA | AB026044.1 | - | 7 | 861 | ||
| AF237906.1 | - | 7 | 849 | |||
| AY039038.1 | - | 7 | 849 | |||
| BC042719.1 | - | 7 | 897 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a8a| Gene Symbol: Ms4a8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022430.2 | NP_071875.2 | 7 | 911 | ||
| GenBank mRNA | AB026044.1 | - | 7 | 861 | ||
| AF237906.1 | - | 7 | 849 | |||
| AY039038.1 | - | 7 | 849 | |||
| BC042719.1 | - | 7 | 897 | |||