siRNA Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166221.1 | NP_001159693.1 | 4 | 308 | ||
| NM_006649.3 | NP_006640.2 | 4 | 308 | |||
| XM_017029237.1 | XP_016884726.1 | 4 | 242 | |||
| XM_017029238.1 | XP_016884727.1 | 4 | 300 | |||
| GenBank mRNA | AK291918.1 | - | 4 | 275 | ||
| AK298578.1 | - | 4 | 277 | |||
| BC001149.1 | - | 4 | 244 | 3355662 | ||
| BC009649.1 | - | 4 | 246 | 3459192 | ||
| BC014987.1 | - | 4 | 236 | 4901477 | ||
| BP353997.1 | - | 4 | 306 | |||
| BU942896.1 | - | 4 | 310 | |||

UTP14A
Genomic Map