siRNA Details
| Gene Symbol: Lpin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199118.1 | NP_001186047.1 | 10, 11 | 1708 | ||
| NM_022883.3 | NP_075021.1 | 10, 11 | 1785 | |||
| GenBank mRNA | AF286724.1 | - | 10, 11 | 1722 | ||
| AK033647.1 | - | 10, 11 | 1706 | |||
| AK049363.1 | - | 10, 11 | 1701 | |||
| AK163833.1 | - | 10, 11 | 1700 | |||
| AK220228.1 | - | 10, 11 | 1785 | |||
| BC026729.1 | - | 10, 11 | 1692 | |||
| BC117882.1 | - | 9, 10 | 1649 | 40096038 | ||
| BC117883.1 | - | 9, 10 | 1649 | 40096041 | ||

Lpin3
Genomic Map