siRNA Details
| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193659.1 | NP_001180588.1 | 4 | 1129 | ||
| NM_001193660.1 | NP_001180589.1 | 5 | 1077 | |||
| NM_001193661.1 | NP_001180590.1 | 4 | 964 | |||
| NM_022890.2 | NP_075028.1 | 4 | 1119 | |||
| GenBank mRNA | AK039672.1 | - | 4 | 1011 | ||
| AK049331.1 | - | 4 | 986 | |||
| AK128999.1 | - | 4 | 998 | |||
| AK132155.1 | - | 4 | 988 | |||
| AK166110.1 | - | 4 | 940 | |||
| BC024057.1 | - | 4 | 1028 | 5100104 | ||

Cldn12
Genomic Map