siRNA Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022985.6 | NP_075361.2 | 8 | 987 | ||
| GenBank mRNA | AJ251508.1 | - | 1691 | |||
| AK002775.1 | - | 8 | 988 | |||
| AK144902.1 | - | 7 | 837 | |||
| BC010683.1 | - | 7 | 848 | 2647905 | ||

Zfand6
Genomic Map