siRNA Details
| Gene Symbol: NEU3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006656.5 | NP_006647.3 | 1, 2 | 260 | ||
| GenBank mRNA | AB008185.2 | - | 2, 3 | 175 | ||
| AK022450.1 | - | 1, 2 | 254 | |||
| AK290442.1 | - | 1, 2 | 222 | |||
| BC136397.1 | - | 2, 3 | 594 | 9020384 | ||
| BC144059.1 | - | 2, 3 | 594 | 9052579 | ||
| CN429425.1 | - | 1, 2 | 170 | |||
| DB240271.1 | - | 1, 2 | 260 | |||
| Y18563.1 | - | 1, 2 | 1031 | |||

NEU3
Genomic Map