siRNA Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168240.1 | NP_001161712.1 | 3 | 1508 | ||
| NM_022986.4 | NP_075362.3 | 4 | 1574 | |||
| GenBank mRNA | AK045436.1 | - | 4 | 1370 | ||
| AK076594.1 | - | 4 | 1382 | |||
| BC046301.1 | - | 4 | 1368 | |||

Irak1bp1
Genomic Map