siRNA Details
Gene Symbol: Zic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022987.3 | NP_075363.1 | 2 | 2238 | ||
GenBank mRNA | AB042155.1 | - | 2 | 2260 | ||
AK006747.1 | - | 2 | 636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022987.3 | NP_075363.1 | 2 | 2238 | ||
GenBank mRNA | AB042155.1 | - | 2 | 2260 | ||
AK006747.1 | - | 2 | 636 |