siRNA Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022988.2 | NP_075364.2 | 6 | 1354 | ||
| GenBank mRNA | AK131631.1 | - | 7 | 1717 | ||
| BC065163.1 | - | 6 | 1354 | 6404016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nif3l1| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022988.2 | NP_075364.2 | 6 | 1354 | ||
| GenBank mRNA | AK131631.1 | - | 7 | 1717 | ||
| BC065163.1 | - | 6 | 1354 | 6404016 | ||