siRNA Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022993.3 | NP_075369.2 | 4 | 688 | ||
| GenBank mRNA | AB042200.1 | - | 4 | 590 | ||
| AK132169.1 | - | 672 | ||||
| AK144035.1 | - | 4 | 701 | |||
| AK170325.1 | - | 4 | 662 | |||
| AK185029.1 | - | 2 | 154 | |||
| AK197457.1 | - | 2 | 128 | |||
| AK203282.1 | - | 2 | 117 | |||
| BC052378.1 | - | 4 | 664 | 5696275 | ||
| BC054447.1 | - | 4 | 656 | 6477698 | ||

Lrp10
Genomic Map