siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 17 | 2591 | ||
GenBank mRNA | AF226323.1 | - | 17 | 2610 | ||
AK077506.1 | - | 1 | 85 | |||
AK135680.1 | - | 17 | 2903 | |||
AK145421.1 | - | 16 | 2560 | |||
AK145853.1 | - | 17 | 2639 | |||
AK150682.1 | - | 17 | 2605 | |||
AK162726.1 | - | 17 | 2570 | |||
AK166821.1 | - | 17 | 2584 | |||
AK167421.1 | - | 17 | 2608 | |||
AK167594.1 | - | 17 | 2604 | |||
AK170473.1 | - | 17 | 2625 | |||
BC005469.1 | - | 17 | 2616 | 3492842 | ||
BC006637.1 | - | 17 | 2596 | 3969440 | ||
U47024.1 | - | 16 | 2561 |
