siRNA Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023047.2 | NP_075534.1 | 4 | 779 | ||
| GenBank mRNA | AF249295.1 | - | 4 | 943 | ||
| AK044866.1 | - | 4 | 651 | |||
| BC065054.1 | - | 4 | 779 | 5709296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpysl5| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023047.2 | NP_075534.1 | 4 | 779 | ||
| GenBank mRNA | AF249295.1 | - | 4 | 943 | ||
| AK044866.1 | - | 4 | 651 | |||
| BC065054.1 | - | 4 | 779 | 5709296 | ||