siRNA Details
| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168218.1 | NP_001161690.1 | 1, 2 | 381 | ||
| NM_001168219.1 | NP_001161691.1 | 1, 2 | 381 | |||
| NM_023197.3 | NP_075686.2 | 1, 2 | 381 | |||
| GenBank mRNA | AF034580.1 | - | 2, 3 | 548 | ||
| AK009230.1 | - | 1, 2 | 42 | |||
| AK083845.1 | - | 1, 2 | 68 | |||
| AK166956.1 | - | 1, 2 | 67 | |||

Knop1
Genomic Map